missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 87-99-0 |
| Physical Form | Neat |
| Linear Formula | HOCH2 [CH(OH)]3 CH2 OH |
| MDL Number | MFCD00064292 |
| Melting Point | 94°C to 97°C |
| Synonym | Xylite |
| Type | Food Additive Standard |
| Quantity | 1 g |
| Formula Weight | 152.15 |
| Grade | Analytical Standard |