missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 99+%

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

Supplier:  Thermo Scientific Chemicals 225980025

 View more versions of this product

Catalog No. AC225980025

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylitol
87-99-0
White
Authentic
Plastic bottle
HOCH2[CH(OH)]3CH2OH
2.5 kg
15, 10283
Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
OCC(O)C(O)C(O)CO
152.15
152.15
Crystalline Powder
99+%
92.0°C to 96.0°C
215.0°C to 217.0°C
2% max. Other polyols (GC)
C5H12O5
MFCD00064291,MFCD00064292
01, 531
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
HEBKCHPVOIAQTA-UHFFFAOYNA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
99+%

Chemical Identifiers

87-99-0
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
6912
OCC(O)C(O)C(O)CO
C5H12O5
MFCD00064291,MFCD00064292
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
(2S,4R)-pentane-1,2,3,4,5-pentol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 201-788-0

RTECSNumber : ZF0800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only