missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylitol, 99%

Catalog No. AAA1694422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1694422 100 g
AAA1694436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1694422 Supplier Thermo Scientific Chemicals Supplier No. A1694422

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

Solubility
Soluble in water (50 mg/ml), abs methanol (6 g/100ml), abs ethanol (1.2 g/100ml), alcohol (very slightly), and pyridine. Insoluble in ether.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas.

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92°C to 96°C
Density 1.515 g/mL
Assay Percent Range 99%
Quantity 100 g
Beilstein 1720523
Sensitivity Hygroscopic
Merck Index 14,10085
Solubility Information Soluble in water (50mg/ml),abs methanol (6g/100ml),abs ethanol (1.2g/100ml),alcohol (very slightly),and pyridine. Insoluble in ether.
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Xylitol
Show More Show Less
EINECSNumber 201-788-0
RTECSNumber ZF0800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only