missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenol Orange Triturate R, 1% (w/w) in Potassium Nitrate, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Xylenol Orange Triturate R |
| Color | Reddish Brown |
| CAS | 7757-79-1, 3618-43-7, 7757-79-1 |
| Boiling Point | 400°C |
| CAS Max % | 1%, 99% |
| Melting Point | 334°C |
| CAS Min % | 99%, 1% |
| Composition | 1% (w/w) in Potassium Nitrate |
| Percent Purity | 1% |
| Grade | Laboratory |
| Show More |