missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, ACS

CAS: 3618-43-7

$102.37 - $1169.92

Chemical Identifiers

CAS 3618-43-7
MDL Number MFCD00044293
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4137904
View Documents
Thermo Scientific Chemicals
04137904
2 g
Each for $102.37
Only null left
 
AA4137909
View Documents
Thermo Scientific Chemicals
04137909
10 g
Each for $334.26
Only null left
 
AA4137918
View Documents
Thermo Scientific Chemicals
04137918
50 g
Each for $1,169.92
Only null left
 
Description

Description

Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3618-43-7
MFCD00044293

Specifications

210°C (decomposition)
MFCD00044293
760.59
Odorless
Powder
3618-43-7
Soluble in water.
ACS
2 g
Xylenol Orange tetrasodium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-805-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only