missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange, TCI America™

Catalog No. B04771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B04771G Supplier TCI America Supplier No. B04771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used as a metal indicator.

Chemical Identifiers

CAS 1611-35-4
Molecular Formula C31H32N2O13S
Molecular Weight (g/mol) 672.658
MDL Number MFCD00014580
InChI Key ORZHVTYKPFFVMG-UHFFFAOYSA-N
Synonym 3,3′C-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonphthalein Disodium Salt
PubChem CID 73041
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4)CN(CC(=O)O)CC(=O)O)O)C)CN(CC(=O)O)CC(=O)O)O

Specifications

Color Brown to Red
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 716.62
Grade Reagent
Chemical Name or Material Xylenol Orange