accessibility menu, dialog, popup

UserName

CH3NOHOOHOOHOSOOCH3OHNHOOHOO

CAS RN 1611-35-4

IUPAC Name: 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Synonyms: xylenol orange
Molecular Weight (g/mol): 672.66
Molecular Formula: C31H32N2O13S
InChi Key: ORZHVTYKPFFVMG-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)