missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol orange, sodium salt, ACS reagent

Catalog No. ac447600050
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC447600050 5 g Glass bottle
AC447600250 25 g Glass bottle
2 options

Catalog No. AC447600050

Supplier: Thermo Scientific Chemicals 447600050

Only null left

CAS: 3618-43-7 | C31H28N2Na4O13S | 760.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
Molecular Weight (g/mol) 760.59

Specifications

Melting Point 210°C
Packaging Glass bottle
Quantity 5 g
Merck Index 15,10281
Solubility Information Solubility in water: 510g/L (20°C)
Formula Weight 760.59
Clarity Passes test
Grade ACS Reagent
Chemical Name or Material Xylenol orange, sodium salt
EINECSNumber 222-805-8

RUO – Research Use Only