missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Disodium Salt, For spectrophotometric determination of metal ions, MilliporeSigma™ Supelco™

Catalog No. 111013282
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013282 1 g
111013283 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013282 Supplier Merck Emd Millipore Supplier No. 520971G

Chemical Identifiers

CAS 1611-35-4
Molecular Formula C31H32N2O13S
Molecular Weight (g/mol) 672.66
InChI Key ORZHVTYKPFFVMG-UHFFFAOYSA-N
Synonym 3,3 '-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt
IUPAC Name 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Linear Formula C31H30N2Na2O13S
Formula Weight 716.62
Quantity 1 g