missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 99%, ACS reagent

CAS: 1330-20-7 | C8H10

Supplier:  Thermo Scientific Chemicals 422680040

 View more versions of this product

Catalog No. AC422680040

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylenes
0.865g/mL
Authentic
Liquid
0.002% max.
C8H10
MFCD00077264
0.05% max. (Coulometric)
15, 10278
106.17
ACS Reagent
1330-20-7,100-41-4
4 L
EcoSafPak™
1.4960 to 1.4980 (20°C, 589nm)
99%
C6H4(CH3)2
Vapor Pressure: 8mbar at 20°C
05, 360
Dimethylbenzene
≥98.5% (xylene isomers plus ethylbenzene (ethylbenzene not to exceed 25%) Separat. techn. GC)

Chemical Identifiers

1330-20-7
C8H10
MFCD00077264
Dimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only