missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 99%, ACS reagent

Catalog No. AC422680200
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422680200 20 L
AC422685000 500 mL
AC422680025 2.5 L
AC422680040 4 L
AC422680250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC422680200 Supplier Thermo Scientific Chemicals Supplier No. 422680200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1330-20-7 | C8H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzene

Specifications

Density 0.865g/mL
Quantity 20 L
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Residue after Evaporation 0.002% max.
Assay Percent Range 99%
Vapor Pressure Vapor Pressure: 8mbar at 20°C
Linear Formula C6H4(CH3)2
Water 0.05% max. (Coulometric)
Beilstein 05, 360
Merck Index 15, 10278
Formula Weight 106.17
Percent Purity ≥98.5% (xylene isomers plus ethylbenzene (ethylbenzene not to exceed 25%) Separat. techn. GC)
Grade ACS
Chemical Name or Material Xylenes
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-535-7
RTECSNumber ZE2100000
TSCA TSCA

RUO – Research Use Only