missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Purified (Dimethylbenzenes), Reagents

Catalog No. p-200021050
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336874 20 L
01336873 4 L
01336872 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336874 Supplier Labchem Inc Supplier No. C234800020E6
Only null left

Xylenes, Purified (Dimethylbenzenes), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym Methyl Toluene, Dimethylbenzenes
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -47.9°C to 13.3°C
Color Colorless
Boiling Point 137°C to 140°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.87
Grade Pure
Chemical Name or Material Xylenes, Purified (Dimethylbenzenes)