missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Purified (Dimethylbenzenes), Reagents

Catalog No. 01336874
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336874 20 L
01336873 4 L
01336872 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336874 Supplier Labchem Inc Supplier No. C234800020E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xylenes, Purified (Dimethylbenzenes), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym Methyl Toluene, Dimethylbenzenes
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -47.9°C to 13.3°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.87
Boiling Point 137°C to 140°C
Grade Pure
Chemical Name or Material Xylenes, Purified (Dimethylbenzenes)