missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, mixed, 97+%

CAS: 1330-20-7 | C8H10

Supplier:  Thermo Scientific Chemicals L13317AP

 View more versions of this product

Catalog No. AAL13317AP


Only null left
Explore available promotions

Description

Description

Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

Solubility
Miscible with aromatic hydrocarbons. Immiscible with water.

Notes
Contains a mixture of the ortho, meta, and para isomers of xylene. May also contain ethylbenzene. Incompatible with strong oxidizing agents.
Specifications

Specifications

Xylenes
mixed
0.86
500 mL
Aromatic
C8H10
UN1307
14,10081
Miscible with aromatic hydrocarbons. Immiscible with water.
≥97%
<-25°C
1330-20-7
137°C to 144°C
29°C (84°F)
1.497
MFCD00077264
1901563
Dimethylbenzenes + ethylbenzene
106.17

Chemical Identifiers

1330-20-7
C8H10
MFCD00077264
Dimethylbenzenes + ethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H312+H332-H315-H319-H335-H373

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only