missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Solstice

Catalog No. 60044941
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044941 1 L Glass Bottle
60044942 2.5 L Glass Bottle
60044945 6 x 1 L Glass Bottle
60044944 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60044941 Supplier Solstice Supplier No. 164461L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for analysis (isomeric mixture), >99.0%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Melting Point -48°C
Color Colorless
Density 0.86g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.4960 to 1.4990
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Boiling Point 145°C
Molecular Weight (g/mol) 106.17
Formula Weight 106.17g/mol
Grade Reagent
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H304-H312 + H332-H315-H319-H335-H373
DOTInformation class 3 - PG 3 - Xylenes
EINECSNumber 215-535-7
RTECSNumber ZE2100000