missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Histology, MilliporeSigma™

Catalog No. MXX00604
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MXX00604 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MXX00604 Supplier Technidata Supplier No. XX00604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality clearing agent for routine histology and cytology applications meeting or exceeding ACS specifications.

Chemical Identifiers

CAS 1330-20-7
Molecular Weight (g/mol) 106.17
Synonym Dimethylbenzenes

Specifications

Quantity 4 L
Packaging Poly Bottle
Grade Histological
Chemical Name or Material Xylenes