missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Histological Grade, Honeywell™

Catalog No. p-7120903
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
4 L
Packaging:
Aluminum Bottle
Catalog No. Quantity Packaging
60047036 4 L Aluminum Bottle
1 options

Catalog No. 60047036

Supplier: Honeywell Inc 5340564L

Histological Grade

  • Commonly used for tissue sampling and processing
  • Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency
  • Available in a variety of packaging options (amber glass bottles, carbon steel flex-spout cans and carbon steel cans with NPT threads)

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Color Colorless
Density 0.86g/mL at 25°C
Boiling Point 140°C
Quantity 4 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Aluminum Bottle
Physical Form Liquid
Refractive Index n20/D 1.497
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Formula Weight 106.17g/mol
Grade Histological
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
DOTInformation class 3 - PG 3 - Xylenes