missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Histological Grade, Solstice

Catalog No. 60047036
Change view
Click to view available options
Quantity:
4 L
Packaging:
Aluminum Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047036 4 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047036 Supplier Honeywell Inc Supplier No. 5340564L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Histological Grade

  • Commonly used for tissue sampling and processing
  • Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency
  • Available in a variety of packaging options (amber glass bottles, carbon steel flex-spout cans and carbon steel cans with NPT threads)

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Color Colorless
Density 0.86g/mL at 25°C
Quantity 4 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Aluminum Bottle
Physical Form Liquid
Refractive Index n20/D 1.497
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Boiling Point 140°C
Molecular Weight (g/mol) 106.17
Formula Weight 106.17g/mol
Percent Purity 75.0%
Grade Histological
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
DOTInformation class 3 - PG 3 - Xylenes