missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, MilliporeSigma™

Catalog No. MXX006020
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MXX006020 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MXX006020 Supplier Technidata Supplier No. XX006020

missing translation for 'validMainframeMsg'

Clearing agent designed for routine histology and cytology applications that meets or exceeds ACS specifications.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C6H4(CH3)2
Molecular Weight (g/mol) 106.17

Specifications

Color Colorless
Quantity 200 L
Packaging Bottle
Physical Form Liquid
Formula Weight 106.17
Grade ACS
Chemical Name or Material Xylenes