missing translation for 'onlineSavingsMsg'
Learn More

Xylenes (Certified ACS), Fisher Chemical™

Catalog No. p-216461
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
X51 1 L Glass Bottle
X54 4 L Glass Bottle
X520 20 L Steel Pail
X5200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. X51 Supplier Thermo Fisher Scientific Supplier No. X5 1
Only null left

Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
MDL Number MFCD00077264

Specifications

Melting Point -34°C
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range Assay: ≥98.5%
Molecular Formula C8H10
Vapor Pressure 8.29mmHg at 25°C
Water 0.05% max.
Synonym Xylol, Dimethylbenzene
Boiling Point 136°C
Formula Weight 106.17
Percent Purity ≥98.5%
Grade Assay: ≥98.5%
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid