missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, ACS Specifications, 98.5% min., ACS, MilliporeSigma™
Specifications
Specifications
| Color | Colorless |
| Quantity | 200 L |
| Packaging | Poly Coated Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | 98.5% min. |
| Formula Weight | 106.17 |
| Percent Purity | ≥98.5% |
| Grade | ACS |
| Chemical Name or Material | Xylenes, For ACS analysis |