missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ACS Reagent (Dimethylbenzenes), Reagents

Catalog No. p-200021119
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337632 200 L
01337631 20 L
01337630 4 L
01337629 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337632 Supplier Labchem Inc Supplier No. C1348000200E
Only null left

Xylenes, ACS Reagent (Dimethylbenzenes), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylenes, ACS Reagent (Dimethylbenzenes)
Melting Point -47.9°C to 13.3°C
CAS 1330-20-7
Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.87
Molecular Formula C6H4(CH3)2
Synonym Methyl Toluene, Dimethylbenzenes
Boiling Point 137°C to 140°C
Molecular Weight (g/mol) 106.165
Grade ACS
Show More Show Less