missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ACS Reagent (Dimethylbenzenes), Reagents

Catalog No. 01337631
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337631 20 L
01337632 200 L
01337630 4 L
01337629 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337631 Supplier Labchem Inc Supplier No. C134800020E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xylenes, ACS Reagent (Dimethylbenzenes), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Xylenes, ACS Reagent (Dimethylbenzenes)
Melting Point -47.9°C to 13.3°C
CAS 1330-20-7
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.87
Molecular Formula C6H4(CH3)2
Synonym Methyl Toluene, Dimethylbenzenes
Boiling Point 137°C to 140°C
Molecular Weight (g/mol) 106.165
Percent Purity 98.5%
Grade ACS
Show More Show Less