Learn More
Xylenes, ACS Grade, ≥ 98.5%, LabChem™
Supplier: LabChem LC2697044
Specifications
| Xylenes | |
| Colorless | |
| Carbon dioxide; Carbon monoxide | |
| 25°C | |
| Amber Glass | |
| ≤0.002% | |
| UN1307 | |
| 106.16 | |
| ACS |
| 1330-20-7 | |
| 100 | |
| 4 x 4 L | |
| Passes Test | |
| Liquid | |
| C8H10 | |
| ≤0.05% | |
| ≥98.5% |
Chemical Identifiers
| 1330-20-7 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Toxic to aquatic life.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
EINECSNumber : 215-535-7
Recommended Storage : Room Temperature