missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ACS Grade, ≥ 98.5%, LabChem™

Catalog No. LC269704
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC269704 4 L Amber Glass
LC2697044 4 x 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC269704 Supplier LabChem Supplier No. LC269704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1330-20-7 | 106.17 g/mol

Chemical Identifiers

CAS 1330-20-7

Specifications

Color Colorless
CAS Max % 100
Decomposition Information Carbon dioxide; Carbon monoxide
Quantity 4 L
Flash Point 25°C
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤0.002%
Linear Formula C8H10
UN Number UN1307
Water ≤0.05%
Molecular Weight (g/mol) 106.16
Formula Weight 106.16
Percent Purity ≥98.5%
Grade ACS
Chemical Name or Material Xylenes
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 215-535-7
Recommended Storage Room Temperature