missing translation for 'onlineSavingsMsg'
Learn More

Xylenes ACS, MilliporeSigma™

Catalog No. p-3176708
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Glass Bottle
Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MXX00556 500 mL Glass Bottle
MXX00553 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MXX00556 Supplier Technidata Supplier No. XX00556
Only null left

Chemical Identifiers

CAS 1330-20-7

Specifications

Quantity 500 mL
Packaging Glass Bottle
Linear Formula C6H4(CH3)2
Formula Weight 106.17
Grade ACS Reagent
Chemical Name or Material Xylenes