missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes ACS, MilliporeSigma™
Specifications
Specifications
| Quantity | 20 L |
| Packaging | Metal Can |
| Linear Formula | C6H4(CH3)2 |
| Molecular Weight (g/mol) | 106.17 |
| Formula Weight | 106.17 |
| Percent Purity | ≥98.5% |
| Grade | ACS |
| Chemical Name or Material | Xylenes |