missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 98+%, Extra Dry, mixed isomers, AcroSeal™

CAS: 1330-20-7 | C8H10

Supplier:  Thermo Scientific Chemicals 396930010

 View more versions of this product

Catalog No. AC396930010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylenes
Mixture of isomers, Extra Dry
Colorless
136.0°C to 140.0°C
23°C
Liquid
98+%
C8H10
MFCD00077264
0.005% Max. (Coulometric)
Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
≥98%
-34.0°C
1330-20-7,100-41-4
0.8650g/mL
1 L
AcroSeal™ Glass Bottle
0.0003% max.
0.865
C6H4(CH3)2
0.6 mPa.s (20°C)
15, 10278
106.17
Extra Dry

Chemical Identifiers

1330-20-7
C8H10
MFCD00077264
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only