missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 98+%, for analysis, mixture of isomers

Catalog No. p-4813715
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC383940010 1 L
AC383940050 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC383940010 Supplier Thermo Scientific Chemicals Supplier No. 383940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1330-20-7 | C8H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
Synonym Dimethylbenzene

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Boiling Point 136.0°C to 140.0°C
Quantity 1 L
Flash Point 23°C
Assay Percent Range 98+%
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Viscosity 0.6 mPa.s (20°C)
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity 98+%
Grade Analytical
Chemical Name or Material Xylenes
Show More Show Less

RUO – Research Use Only