missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, 98.5%, MilliporeSigma™
Specifications
Specifications
| Quantity | 4 L |
| Flash Point | 29.4°C |
| Packaging | Glass Bottle (Pre-packed) |
| Residue after Evaporation | 0.002% max |
| Assay Percent Range | 98.5% min |
| Percent Purity | ≥98.5% (GC) |
| Grade | ACS |
| Chemical Name or Material | Xylene |
Safety and Handling
| Recommended Storage | Store at room temperature |