missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ≥98.5%, ACS Reagent, xylenes plus ethylbenzene basis, Honeywell™ Riedel-de Haën™

Catalog No. p-7120902
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045658 1 L Glass Bottle
60045659 2.5 L Glass Bottle
60045660 20 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045658 Supplier Honeywell Chemicals Supplier No. 2476421LCB
Only null left

ACS Reagent,≥98.5% xylenes + ethylbenzene basis

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264

Specifications

Melting Point -48°C
Density 0.86g/mL at 25°C
Boiling Point 137°C to 140°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Formula Weight 106.17g/mol
Percent Purity ≥98.5% xylenes plus ethylbenzene basis
Grade ACS Reagent
Chemical Name or Material Xylenes
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H304-H312 + H332-H315-H319-H335-H373-H412
DOTInformation Class 3 - PG 3 - Xylenes