missing translation for 'onlineSavingsMsg'
Learn More

Xylenecyanol FF, Electrophoresis Reagent, dye content 70%

Catalog No. p-7076670
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6637709 10 g Glass bottle
AAJ6637718 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6637709 Supplier Thermo Scientific Chemicals Supplier No. J6637709
Only null left

CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Molecular Weight (g/mol) 538.61
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
Synonym Acid Blue 147,C.I. 42135
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point ∼295°C (decomposition)
Packaging Glass bottle
Quantity 10 g
Formula Weight 538.61
Percent Purity dye content 70%
Physical Form Powder
Chemical Name or Material Xylenecyanol FF
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-167-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only