missing translation for 'onlineSavingsMsg'
Learn More

Xylenecyanol FF, dye content 70%

Catalog No. p-7025836
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2153009 10 g
AAB2153018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2153009 Supplier Thermo Scientific Chemicals Supplier No. B2153009
Only null left

CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol

Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.

Chemical Identifiers

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Molecular Weight (g/mol) 538.61
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
Synonym Acid blue 147; C.I. 42135
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point ∼295°C (decomposition)
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 538.61
Chemical Name or Material Xylenecyanol FF, dye content 70%
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-167-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only