missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylazine Hydrochloride 98.0+%, TCI America™
$186.41 - $562.95
Chemical Identifiers
| CAS | 23076-35-9 |
|---|---|
| Molecular Formula | C12H17ClN2S |
| Molecular Weight (g/mol) | 256.79 |
| MDL Number | MFCD00058196 |
| InChI Key | QYEFBJRXKKSABU-UHFFFAOYSA-N |
| Synonym | xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470 |
| PubChem CID | 68554 |
| IUPAC Name | hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride |
| SMILES | [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1 |
Chemical Identifiers
| 23076-35-9 | |
| 256.79 | |
| QYEFBJRXKKSABU-UHFFFAOYSA-N | |
| 68554 | |
| [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1 |
| C12H17ClN2S | |
| MFCD00058196 | |
| xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470 | |
| hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride |
Specifications
| 23076-35-9 | |
| 5 g | |
| C12H17ClN2S | |
| 2811 | |
| QYEFBJRXKKSABU-UHFFFAOYSA-N | |
| hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride | |
| 68554 | |
| ≥98.0% (HPLC,T) |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00058196 | |
| xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470 | |
| [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1 | |
| 256.79 | |
| 256.79 | |
| Xylazine Hydrochloride |
Safety and Handling
RTECSNumber : XJ0776350
TSCA : No