missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529359
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (90%), Rat (90%), Dog (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.Chemical Identifiers
| XRCC1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| XRCC1 | |
| DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1 | |
| XRCC1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.73 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P18887, Q9ESZ0 | |
| 7515, 84495 | |
| Recombinant protein encompassing a sequence within the center region of human XRCC1. The exact sequence is proprietary. | |
| Antibody |
Specifications
| XRCC1 | |
| Polyclonal | |
| Unconjugated | |
| XRCC1 | |
| DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7515, 84495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P18887, Q9ESZ0 | |
| XRCC1 | |
| Recombinant protein encompassing a sequence within the center region of human XRCC1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |