missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPR1 Recombinant Rabbit Monoclonal Antibody (HL2632)
GREENER_CHOICE

Catalog No. MA556923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556923 Supplier Invitrogen™ Supplier No. MA556923
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

XPR1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.

Specifications

Antigen XPR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2632
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Xpr1
Gene Accession No. Q9UBH6, Q9Z0U0
Gene Alias IBGC6; Protein SYG1 homolog; receptor for MCF virus 1; Rmc1; Rmc-1; RP11-46A10.1; suppressor of yeast G>a> deletion; susceptibility to xenotropic virus; Sxv; Syg1; X3; xenotropic and polytropic murine leukemia virus receptor; xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog; XPR1; XR; X-receptor
Gene Symbols Xpr1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human XPR1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19775, 289424, 9213
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less