missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ XPR Excellence Automatic Balance

Catalog No. 15467608
Change view
Click to view available options
Capacity:
121 g, 320 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
15467608 121 g, 320 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15467608 Supplier METTLER TOLEDO™ Supplier No. 30957900

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automatically dispense powders for highly accurate weighing of very small samples

  • Liquid is an additional accessory
  • Adjustment: Internal (automatic/FACT)
  • Automatic static detection
  • Automatic doors
  • Leveling guidance
  • Automated dispensing
  • Supports 21 CFR Part 11 (LabX compatible)
  • Includes EQPac Installation and Qualification to support the highest level of regulatory compliance
  • Provides regulatory conformance qualification documentation and professional qualification including test procedures and calibration
  • Installation is completed by a factory-trained METTLER TOLEDO Service Technician

Specifications

Connection Type Ethernet (LAN), USB, RS232, Bluetooth (optional)
Capacity 121 g, 320 g
Display Type Color Touchscreen
Readability 0.01 mg, 0.1 mg
Material Die-Cast Aluminum, Plastic, Chrome Steel and Glass
Min. Sample Weight 40 mg
Model XPR305DRQ
Stabilization Time 1.5 seconds
Type Automatic Balance
Electrical Requirements Input: 100/240 V, AC plusmn;10%, 50/60 Hz, 1.8 A
Legal for Trade No
Show More Show Less