missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPOT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566095 Supplier Invitrogen™ Supplier No. PA566095
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: MDEQALLGLNP NADSDFRQRA LAYFEQLKIS PDAWQVCAEA LAQRTYSDDH VKFFCFQVLE HQVKYKYSEL TTVQQQLIRE TLISWLQAQM LNPQP Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.

Specifications

Antigen XPOT
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XPOT
Gene Accession No. O43592
Gene Alias 1110004L07Rik; 3110065H13Rik; AI452076; C79645; exportin for tRNA; exportin(tRNA); exportin, tRNA; exportin, tRNA (nuclear export receptor for tRNAs); exportin-T; tRNA exportin; XPO3; XPOT
Gene Symbols XPOT
Host Species Rabbit
Immunogen Recombinant Human XPOT. Recombinant protein control fragment (Product #RP-105080).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less