missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675246
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01675246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01675246 Supplier Invitrogen™ Supplier No. PA141633
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

PA1-41633 detects Exportin-4 in chicken, chimpanzee, rhesus monkey, canine, human, rat and mouse samples. This antibody is predicted to react with bovine, xenopus, and zebrafish based on sequence homology. Suggested positive control: Jurkat whole cell lysate. The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c), rhesus monkey, chimp (isoforms 1-6), 94% homologous in bovine and xenopus, and 88% homologous in zebrafish.

Nuclear transport receptors of karyopherin/importin-beta superfamily mediate various transport pathways between nucleus and cytoplasm. Exportin-4 is a novel mammalian member of this family. It is a RanGTP-binding protein consisting of N-terminal RanGTP-binding motif. It functions as a nuclear export receptor for eIF-5A and also might function as an export adapter in RNA export. It is also involved in nuclear export of Smad3 and regulation of Smad signaling.

Specifications

Antigen XPO4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene XPO4
Gene Accession No. Q5ZMR9, Q9C0E2, Q9ESJ0
Gene Alias B430309A01Rik; exp4; exportin 4; exportin-4; FLJ13046; hypothetical protein; KIAA1721; mKIAA1721; RCJMB04_1f17; Xpo4
Gene Symbols XPO4
Host Species Rabbit
Immunogen A portion of amino acids 850-950 of human Exportin-4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290280, 428081, 477344, 57258, 64328
Target Species Human, Mouse, Rat, Canine, Monkey, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less