missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ xPIPETTE Ergonomic 12-Channel Pipettes

Catalog No. p-7185223
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12111406 0.5 to 10 μL
12111407 2 to 20 μL
12111408 20 to 200 μL
12111409 20 to 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12111406 Supplier BIOTIX™ Supplier No. 30537451
Only null left

Biotix™ xPIPETTE Ergonomic 12-Channel Pipettes utilize patented ejection technology to reduce fatigue, making it the top choice for improving laboratory ergonomics. Designed with durable and lightweight components, xPIPETTE produces precise and accurate liquid transfer while maintaining hand comfort.

  • The xPIPETTE Multi-channel is part of the Biotix Ergonomix pipetting system
  • Features positive stop technology to reduce fatigue by drastically lowering ejection forces when compared to universal models
  • That means less hand and wrist force is required from every tip, leading to more accurate and precise measurements even after a long day of pipetting
  • Calibrated to ISO 8655 standards with the Biotix xTIP4
  • Fully compatible with Rainin LTS tips
  • Accurate and precise with low channel to channel variability for reproducible results
  • Lightweight components and spring forces produce a smooth, controlled aspiration with perfect balance
  • The padded ejector button (symmetrically designed for left or right hand users) reduces the dynamic forces and vibrations during ejection

Specifications

Certifications/Compliance ISO 8655
Compatible Tips Biotix xTIP4, Rainin LTS
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With Pipette Tips
Grip Type Ergonomic
Material PVDF
Model Biotix xPIPETTE
Number of Channels 12
Product Line xPIPETTE
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
Show More Show Less