missing translation for 'onlineSavingsMsg'
Learn More

Xenon difluoride, 99.5% (metals basis)

Catalog No. AA3973904
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3973904 2 g
AA3973909 10 g
AA3973918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3973904 Supplier Thermo Scientific Chemicals Supplier No. 03973904

Fluorinating agent | CAS: 13709-36-9 | F2Xe | 169.29 g/mol

Used as Fluorinating agent and oxidizing agent, gaseous etchant for silicon, particularly in the production of microelectromechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13709-36-9
Molecular Formula F2Xe
Molecular Weight (g/mol) 169.29
MDL Number MFCD00040538
InChI Key IGELFKKMDLGCJO-UHFFFAOYSA-N
Synonym xenon difluoride, xenon fluoride, xenon fluoride xef2, unii-6poj14981p, bis fluoranyl xenon, xef2, acmc-20aj9j, xenon difluoride trace metals basis
PubChem CID 83674
IUPAC Name difluoroxenon
SMILES F[Xe]F

Specifications

Melting Point 127°C to 129°C
Color White
Physical Form Crystalline Granules
Quantity 2 g
UN Number UN3087
Merck Index 14,10072
Solubility Information Decomposes in water.
Formula Weight 169.3
Percent Purity 99.5%
Sensitivity Moisture sensitive
Density 4.32
Assay (metals basis)
Chemical Name or Material Xenon difluoride
Show More Show Less
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H272-H301-H330-H314-H318;
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P380+P375-P372-P380-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
EINECSNumber 237-251-2
RTECSNumber ZE1294116
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only