missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthurenic Acid 96.0+%, TCI America™
$215.62 - $215.62
Chemical Identifiers
| CAS | 59-00-7 |
|---|---|
| Molecular Formula | C10H7NO4 |
| Molecular Weight (g/mol) | 205.17 |
| MDL Number | MFCD00006754 |
| InChI Key | FBZONXHGGPHHIY-UHFFFAOYSA-N |
| Synonym | xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429 |
| PubChem CID | 5699 |
| ChEBI | CHEBI:10072 |
| IUPAC Name | 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid |
| SMILES | OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1 |
Chemical Identifiers
| 59-00-7 | |
| 205.17 | |
| FBZONXHGGPHHIY-UHFFFAOYSA-N | |
| 5699 | |
| 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid |
| C10H7NO4 | |
| MFCD00006754 | |
| xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429 | |
| CHEBI:10072 | |
| OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1 |
Specifications
| 59-00-7 | |
| MFCD00006754 | |
| xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429 | |
| OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1 | |
| 205.17 | |
| CHEBI:10072 | |
| ≥96.0% (HPLC,T) | |
| Xanthurenic Acid |
| C10H7NO4 | |
| 1 g | |
| FBZONXHGGPHHIY-UHFFFAOYSA-N | |
| 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid | |
| 5699 | |
| 205.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UZ9275000
TSCA : No