accessibility menu, dialog, popup

UserName

CAS RN 59-00-7

IUPAC Name: 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
Synonyms: xanthurenic acid
Molecular Weight (g/mol): 205.17
Molecular Formula: C10H7NO4
InChi Key: FBZONXHGGPHHIY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)