missing translation for 'onlineSavingsMsg'
Learn More

Xanthurenic Acid 96.0+%, TCI America™

Catalog No. X00541G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X00541G Supplier TCI America Supplier No. X00541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-00-7
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.17
MDL Number MFCD00006754
InChI Key FBZONXHGGPHHIY-UHFFFAOYSA-N
Synonym xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
PubChem CID 5699
ChEBI CHEBI:10072
IUPAC Name 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1

Specifications

Quantity 1 g
Formula Weight 205.17
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Xanthurenic Acid
RTECSNumber UZ9275000
TSCA No