missing translation for 'onlineSavingsMsg'
Learn More

Xanthotoxin, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013845
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013845 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013845 Supplier Merck Emd Millipore Supplier No. 5644850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Xanthotoxin is an allelochemical or a phototoxic allomone mainly found in host plants of black swallowtail. It can have the tendency to induce tolerance towards α-cypermethrin, a pyrethroid insecticide used for insects like Helicoverpa zea.

Specifications

CAS 298-81-7
Physical Form Neat
Linear Formula C12H8O4
MDL Number MFCD00005009
Melting Point 148°C to 150°C
Synonym 8-Methoxypsoralen; 8-MOP; 9-Methoxyfuro[3,2-g][1 ]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
Type Analytical Standard
Quantity 50 mg
Formula Weight 216.19
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less