missing translation for 'onlineSavingsMsg'
Learn More

Xanthine 98.0+%, TCI America™

Supplier:  TCI America X000425G

Catalog No. X000425G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Xanthine
C5H4N4O2
25 g
LRFVTYWOQMYALW-UHFFFAOYSA-N
3,7-dihydropurine-2,6-dione
1188
152.11
Crystalline Powder
69-89-6
MFCD00078453
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
C1=NC2=C(N1)C(=O)NC(=O)N2
152.113
CHEBI:17712
≥98.0% (HPLC)

Chemical Identifiers

69-89-6
152.113
LRFVTYWOQMYALW-UHFFFAOYSA-N
1188
3,7-dihydropurine-2,6-dione
C5H4N4O2
MFCD00078453
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
CHEBI:17712
C1=NC2=C(N1)C(=O)NC(=O)N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1133

RTECSNumber : ZD7700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions