accessibility menu, dialog, popup

UserName

CAS RN 69-89-6

IUPAC Name: 2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: xanthine
Molecular Weight (g/mol): 152.11
Molecular Formula: C5H4N4O2
InChi Key: LRFVTYWOQMYALW-UHFFFAOYSA-N
SMILES: O=C1NC2=C(NC=N2)C(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)