missing translation for 'onlineSavingsMsg'
Learn More

Xanthene-9-carboxylic Acid 98.0+%, TCI America™

Catalog No. X00315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00315G 5 g
X003125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00315G Supplier TCI America Supplier No. X00315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82-07-5
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00005059
InChI Key VSBFNCXKYIEYIS-UHFFFAOYSA-N
Synonym xanthene-9-carboxylic acid, xanthanoic acid, xanthenecarboxylic acid, 9-xanthenecarboxylic acid, unii-63a81f6a8v, xanthene-9-carboxylicacid, pubchem17697, maybridge1_006340, 9-xanthenylcarboxylic acid, chembl520
PubChem CID 65736
IUPAC Name 9H-xanthene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)C(=O)O

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Xanthene-9-carboxylic Acid