missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthane Hydride 95.0+%, TCI America™
Supplier: TCI America X0001100G
Specifications
| Xanthane Hydride | |
| C2H2N2S3 | |
| 100 g | |
| YWZHEXZIISFIDA-UHFFFAOYSA-N | |
| 5-amino-1,2,4-dithiazole-3-thione | |
| 2735329 | |
| ≥95.0% (HPLC) |
| 6846-35-1 | |
| MFCD00051660 | |
| xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol | |
| C1(=NC(=S)SS1)N | |
| 150.232 | |
| 150.23 | |
| Crystalline Powder |
Chemical Identifiers
| 6846-35-1 | |
| 150.232 | |
| YWZHEXZIISFIDA-UHFFFAOYSA-N | |
| 2735329 | |
| C1(=NC(=S)SS1)N |
| C2H2N2S3 | |
| MFCD00051660 | |
| xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol | |
| 5-amino-1,2,4-dithiazole-3-thione |
Safety and Handling
RTECSNumber : JO6500000
TSCA : No