missing translation for 'onlineSavingsMsg'
Learn More

Xanthane Hydride 95.0+%, TCI America™

Catalog No. p-7137949
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X000125G 25 g
X0001100G 100 g
X0001500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. X000125G Supplier TCI America Supplier No. X000125G
Only null left

Chemical Identifiers

CAS 6846-35-1
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.232
MDL Number MFCD00051660
InChI Key YWZHEXZIISFIDA-UHFFFAOYSA-N
Synonym xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol
PubChem CID 2735329
IUPAC Name 5-amino-1,2,4-dithiazole-3-thione
SMILES C1(=NC(=S)SS1)N

Specifications

Quantity 25 g
Formula Weight 150.23
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Xanthane Hydride
RTECSNumber JO6500000
TSCA No